siRNA Details
Gene Symbol: Pxt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153390.1 | NP_700439.1 | 3 | 448 | ||
GenBank mRNA | AF486826.1 | - | 3 | 448 | ||
AK005593.1 | - | 3 | 451 | |||
BC100437.1 | - | 3 | 447 | 30915792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
