siRNA Details
Gene Symbol: Pacrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027032.2 | NP_081308.1 | 2 | 528 | ||
GenBank mRNA | AK005771.1 | - | 2 | 524 | ||
AK135510.1 | - | 2 | 530 | |||
BC120740.1 | - | 2 | 506 | 40129663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
