siRNA Details
Gene Symbol: 1700001L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027035.1 | NP_081311.1 | 2 | 332 | ||
GenBank mRNA | AK005609.1 | - | 2 | 333 | ||
AK138387.1 | - | 2 | 333 | |||
BC049693.1 | - | 3 | 242 | |||
BC119515.1 | - | 2 | 319 | 40055598 | ||
BC119516.1 | - | 2 | 318 | 40055603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
