siRNA Details
Gene Symbol: 1700012B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162428.1 | NP_001155900.1 | 3 | 456 | ||
NM_027038.1 | NP_081314.1 | 3 | 456 | |||
GenBank mRNA | AK005888.1 | - | 3 | 450 | ||
AK132924.1 | - | 457 | ||||
AK160153.1 | - | 3 | 457 | |||
BC048675.1 | - | 3 | 432 | 6774204 | ||
BC099559.1 | - | 3 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
