siRNA Details
Gene Symbol: 1700012B09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029306.3 | NP_083582.2 | 2 | 217 | ||
GenBank mRNA | AK005747.1 | - | 2 | 218 | ||
AK005890.1 | - | 2 | 200 | |||
BC092535.1 | - | 2 | 179 | 6773440 | ||
CF105954.1 | - | 2 | 166 |
