siRNA Details
Gene Symbol: 1700012B09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029306.3 | NP_083582.2 | 2 | 234 | ||
GenBank mRNA | AK005747.1 | - | 2 | 235 | ||
AK005890.1 | - | 2 | 217 | |||
BC092535.1 | - | 2 | 196 | 6773440 | ||
CF105954.1 | - | 2 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
