siRNA Details
Gene Symbol: 1700007K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027040.1 | NP_081316.1 | 2 | 317 | ||
GenBank mRNA | AK005731.1 | - | 2 | 327 | ||
AY927869.1 | - | 2 | 262 | |||
BC048581.1 | - | 2 | 298 | |||
BC099566.1 | - | 2 | 317 | 30916995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
