siRNA Details
Gene Symbol: 1700003M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027041.4 | NP_081317.4 | 6 | 716 | ||
GenBank mRNA | BC058220.1 | - | 6 | 688 | 6306249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
