siRNA Details
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178617.4 | NP_848732.3 | 11 | 1094 | ||
GenBank mRNA | AK039240.1 | - | 11 | 910 | ||
AK050307.1 | - | 12 | 991 | |||
AK081951.1 | - | 11 | 986 | |||
AY278200.2 | - | 11 | 1077 | |||
BC067055.1 | - | 11 | 1094 | 30545678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
