siRNA Details
Gene Symbol: 1700084M14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006997.1 | - | 2 | 346 | ||
BC048745.1 | - | 2 | 387 | |||
BC100410.1 | - | 2 | 345 |
Gene Symbol: 1700003E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005630.1 | - | 2 | 346 | ||
BC089468.1 | - | 2 | 347 | 6775214 |
Gene Symbol: BC061195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC061195.1 | - | 413 | 6706856 |
