siRNA Details
Gene Symbol: Lrrc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162973.1 | NP_001156445.1 | 2 | 201 | ||
NM_001162974.1 | NP_001156446.1 | 1 | 107 | |||
NM_027053.1 | NP_081329.1 | 3 | 264 | |||
GenBank mRNA | AK005758.1 | - | 3 | 215 | ||
BC025128.1 | - | 1 | 158 | 4225298 | ||
EU627089.1 | - | 1 | 65 | |||
EU627090.1 | - | 1 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
