siRNA Details
Gene Symbol: Cypt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173367.2 | NP_775543.2 | 1 | 179 | ||
GenBank mRNA | AF463500.1 | - | 1 | 159 | ||
AK005674.1 | - | 1 | 182 | |||
AM040452.1 | - | 1 | 174 | |||
BC048527.1 | - | 1 | 179 | 6772990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
