siRNA Details
Gene Symbol: Cst12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027054.1 | NP_081330.1 | 1, 2 | 237 | ||
GenBank mRNA | AF440737.2 | - | 1, 2 | 219 | ||
AK005670.1 | - | 1, 2 | 228 | |||
BC119432.2 | - | 1, 2 | 237 | 40041153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
