siRNA Details
Gene Symbol: 1700012P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027056.1 | NP_081332.1 | 4 | 656 | ||
GenBank mRNA | AK005933.1 | - | 4 | 657 | ||
BC099569.1 | - | 4 | 656 | 30918007 | ||
BC116372.1 | - | 4 | 582 | 40100233 | ||
BC126971.1 | - | 4 | 582 | 40100231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
