siRNA Details
Gene Symbol: Glrx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038592.1 | NP_001033681.1 | 3 | 283 | ||
NM_001038593.1 | NP_001033682.1 | 3 | 186 | |||
NM_001038594.1 | NP_001033683.1 | 3 | 125 | |||
NM_023505.2 | NP_075994.2 | 3 | 162 | |||
GenBank mRNA | AF276918.1 | - | 3 | 129 | ||
AF380337.1 | - | 3 | 226 | |||
AK005853.1 | - | 3 | 186 | |||
AK029540.1 | - | 1 | 1659 | |||
AK142654.1 | - | 3 | 126 | |||
BC065387.1 | - | 3 | 160 | |||
BC132031.1 | - | 3 | 205 | 40130308 | ||
BC132033.1 | - | 3 | 205 | 40130310 | ||
BC145303.1 | - | 3 | 248 | |||
BC145304.1 | - | 3 | 205 | 9053858 | ||
BY226480.1 | - | 3 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
