siRNA Details
Gene Symbol: Mocs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160330.1 | NP_001153802.1 | 1 | 1136 | ||
GenBank mRNA | AK006163.1 | - | 1 | 514 | ||
BC115985.1 | - | 2 | 702 | |||
BE133066.1 | - | 1 | 440 | |||
BG083535.2 | - | 1 | 35 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
