siRNA Details
Gene Symbol: C8g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271777.1 | NP_001258706.1 | 4 | 430 | ||
NM_027062.2 | NP_081338.1 | 4 | 651 | |||
GenBank mRNA | AK005962.1 | - | 4 | 434 | ||
BC019967.1 | - | 4 | 651 | 4023907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
