siRNA Details
Gene Symbol: 1700024P04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027064.1 | NP_081340.1 | 1 | 159 | ||
GenBank mRNA | AK006311.1 | - | 1 | 160 |

More Information
Other Species Targeted

Gene Ontology Categories:
