siRNA Details
Gene Symbol: H2bfm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027067.2 | NP_081343.1 | 1 | 445 | ||
GenBank mRNA | AK005982.1 | - | 1 | 446 | ||
BC117076.1 | - | 1 | 436 | 40126395 | ||
BC117080.1 | - | 1 | 436 | 40126399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
