siRNA Details
Gene Symbol: 1700019A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027070.1 | NP_081346.1 | 6 | 333 | ||
GenBank mRNA | AK006101.1 | - | 6 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
