siRNA Details
Gene Symbol: Plac8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027072.1 | NP_081348.1 | 4 | 571 | ||
GenBank mRNA | AK006002.1 | - | 4 | 549 | ||
BC110558.1 | - | 4 | 533 | 40049443 | ||
BC110559.1 | - | 4 | 533 | 40049451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
