siRNA Details
Gene Symbol: 1700025F22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027074.3 | NP_081350.1 | 5 | 364 | ||
GenBank mRNA | AK006335.1 | - | 5 | 365 | ||
BC048529.1 | - | 4 | 292 | 6774736 | ||
BC061241.1 | - | 5 | 372 | 6743876 |
