siRNA Details
Gene Symbol: 1700027A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006406.1 | - | 3 | 227 | ||
BC048669.1 | - | 2 | 275 | |||
BC100504.1 | - | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
