siRNA Details
Gene Symbol: Clic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027085.3 | NP_081361.1 | 6 | 680 | ||
GenBank mRNA | AK009020.1 | - | 6 | 682 | ||
BC060967.1 | - | 6 | 685 | 30297308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
