siRNA Details
Gene Symbol: Rwdd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145968.1 | NP_001139440.1 | 3 | 609 | ||
NM_001145969.1 | NP_001139441.1 | 3 | 643 | |||
NM_027100.2 | NP_081376.2 | 3 | 738 | |||
GenBank mRNA | AK006533.1 | - | 3 | 737 | ||
BC115691.1 | - | 3 | 738 | 40104313 | ||
BC115692.1 | - | 3 | 609 | 40104314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
