siRNA Details
Gene Symbol: Esam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027102.3 | NP_081378.1 | 4, 5 | 759 | ||
GenBank mRNA | AF361882.1 | - | 4, 5 | 744 | ||
AK009223.1 | - | 4, 5 | 734 | |||
AK156399.1 | - | 4, 5 | 757 | |||
BC019460.1 | - | 4, 5 | 743 | 4188050 | ||
BC049249.1 | - | 4, 5 | 739 | 5039892 | ||
BG077536.2 | - | 2, 3 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
