siRNA Details
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023514.4 | NP_076003.3 | 9 | 934 | ||
GenBank mRNA | AJ250193.1 | - | 5 | 403 | ||
AK009082.1 | - | 9 | 884 | |||
AK049465.1 | - | 8 | 1923 | |||
AK144656.1 | - | 9 | 892 | |||
AK150985.1 | - | 9 | 891 | |||
BC022587.1 | - | 4 | 372 | |||
BC051936.1 | - | 8 | 627 | |||
BC069980.1 | - | 7 | 528 | |||
BC116437.2 | - | 9 | 884 | 40104493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
