siRNA Details
Gene Symbol: Avpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027106.4 | NP_081382.1 | 3 | 877 | ||
GenBank mRNA | AK009243.1 | - | 3 | 878 | ||
AY505175.1 | - | 1 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
