siRNA Details
Gene Symbol: Lyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027111.3 | NP_081387.1 | 6 | 557 | ||
GenBank mRNA | AK009014.1 | - | 6 | 558 | ||
BC119394.1 | - | 6 | 550 | 8734143 | ||
BC119396.1 | - | 6 | 550 | 8734145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
