siRNA Details
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081058.2 | NP_001074527.1 | 11 | 3365 | ||
NM_027118.1 | NP_081394.1 | 11 | 3365 | |||
GenBank mRNA | AK035493.1 | - | 11 | 2036 | ||
AK173265.1 | - | 11 | 3365 | |||
BC051093.1 | - | 4 | 249 | |||
BC076619.1 | - | 12 | 1820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
