siRNA Details
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081058.2 | NP_001074527.1 | 14 | 4401 | ||
NM_027118.1 | NP_081394.1 | 14 | 4221 | |||
GenBank mRNA | AK009373.1 | - | 1 | 286 | ||
AK035493.1 | - | 14 | 3072 | |||
AK148986.1 | - | 1 | 21 | |||
AK173265.1 | - | 14 | 4221 | |||
BC032179.1 | - | 2 | 504 | |||
BC051093.1 | - | 7 | 1105 | |||
BC076619.1 | - | 15 | 2676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
