siRNA Details
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181588.3 | NP_853619.1 | 6 | 638 | ||
GenBank mRNA | AK009374.1 | - | 5 | 630 | ||
BC024580.1 | - | 6 | 662 | 5064316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
