siRNA Details
Gene Symbol: Fastkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027123.4 | NP_081399.3 | 2 | 1093 | ||
GenBank mRNA | AK031142.1 | - | 2 | 1080 | ||
AK032530.1 | - | 2 | 1079 | |||
AK033189.1 | - | 2 | 1094 | |||
AK036778.1 | - | 2 | 1079 | |||
AK148902.1 | - | 2 | 1083 | |||
BC042506.1 | - | 2 | 1070 | 4023412 | ||
BC060688.1 | - | 2 | 1313 | 6409274 |
