siRNA Details
Gene Symbol: Pcgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172716.4 | NP_766304.2 | 6 | 533 | ||
GenBank mRNA | AK033609.1 | - | 6 | 608 | ||
AK085574.1 | - | 6 | 589 | |||
AK089091.1 | - | 6 | 534 | |||
BC049266.1 | - | 400 | 5371040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
