siRNA Details
Gene Symbol: Odam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027128.2 | NP_081404.2 | 9 | 702 | ||
GenBank mRNA | AK009298.1 | - | 6 | 406 | ||
BC132394.1 | - | 6 | 346 | 40130671 | ||
BC132396.1 | - | 6 | 345 | 40130673 | ||
EF113909.1 | - | 9 | 702 |
