siRNA Details
Gene Symbol: Ap5s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291031.1 | NP_001277960.1 | 3 | 988 | ||
NM_027129.3 | NP_081405.2 | 4 | 899 | |||
GenBank mRNA | AK002807.1 | - | 3 | 908 | ||
BC048399.1 | - | 4 | 819 | 6438951 | ||
DV061712.1 | - | 2 | 810 |
