siRNA Details
Gene Symbol: Lce1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027137.2 | NP_081413.1 | 2 | 605 | ||
GenBank mRNA | AK009666.1 | - | 2 | 606 | ||
BC060282.1 | - | 2 | 577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
