siRNA Details
Gene Symbol: Kansl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289437.1 | NP_001276366.1 | 7, 8 | 1356 | ||
NM_001289438.1 | NP_001276367.1 | 7, 8 | 1448 | |||
NM_001289439.1 | NP_001276368.1 | 6, 7 | 1241 | |||
NM_001289440.1 | NP_001276369.1 | 6, 7 | 902 | |||
NM_133714.5 | NP_598475.2 | 7, 8 | 1356 | |||
GenBank mRNA | AK046649.1 | - | 7, 8 | 1326 | ||
AK046983.1 | - | 3, 4 | 295 | |||
AK047269.1 | - | 3, 4 | 294 | |||
AK089025.1 | - | 6, 7 | 1230 | |||
AK150748.1 | - | 7, 8 | 1350 | |||
AK151604.1 | - | 7, 8 | 1351 | |||
AK152604.1 | - | 7, 8 | 1350 | |||
AK153007.1 | - | 7, 8 | 1350 | |||
AK162714.1 | - | 7, 8 | 1329 | |||
AK167429.1 | - | 7, 8 | 1331 | |||
AK168499.1 | - | 6, 7 | 1219 | |||
AK183493.1 | - | 3, 4 | 297 | |||
AK188822.1 | - | 3, 4 | 296 | |||
AK205712.1 | - | 3, 4 | 297 | |||
AK218567.1 | - | 3, 4 | 315 | |||
BC003301.1 | - | 7, 8 | 1332 | 3584464 | ||
BC011085.1 | - | 6, 7 | 897 | |||
BC083325.1 | - | 7, 8 | 1072 | 6828438 | ||
BC092275.1 | - | 5, 6 | 668 | 3492339 | ||
BC125631.1 | - | 7, 8 | 1308 | 40130146 | ||
BC125633.1 | - | 7, 8 | 1308 | 40130148 | ||
BC145047.1 | - | 7, 8 | 1400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
