siRNA Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029556.3 | NP_083832.2 | 2, 3 | 283 | ||
GenBank mRNA | AF428254.1 | - | 2, 3 | 256 | ||
AK002741.1 | - | 2, 3 | 409 | |||
AK009345.1 | - | 2, 3 | 271 | |||
BC023398.1 | - | 2, 3 | 273 | 5064709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
