siRNA Details
Gene Symbol: Cd164l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027152.1 | NP_081428.1 | 3 | 445 | ||
GenBank mRNA | AK009888.1 | - | 3 | 371 | ||
AK139809.1 | - | 3 | 446 | |||
BC111824.1 | - | 3 | 368 | 40055637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
