siRNA Details
Gene Symbol: 2310061I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033630.1 | NP_001028802.1 | 5, 6 | 958 | ||
GenBank mRNA | AK010000.1 | - | 2, 3 | 254 | ||
AK038622.1 | - | 4, 5 | 1810 | |||
AK145087.1 | - | 5, 6 | 912 | |||
AK172070.1 | - | 5, 6 | 875 | |||
BC058418.1 | - | 2, 3 | 203 | |||
BC100355.1 | - | 4, 5 | 587 | 30527188 | ||
BC117035.1 | - | 4, 5 | 618 | |||
BC117039.1 | - | 4, 5 | 618 | |||
BQ043979.1 | - | 2, 3 | 205 | |||
BQ444983.1 | - | 4, 5 | 401 |

More Information
Other Species Targeted

Gene Ontology Categories:
