siRNA Details
Gene Symbol: 2310061I04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033630.1 | NP_001028802.1 | 3 | 631 | ||
GenBank mRNA | AK038622.1 | - | 2 | 1483 | ||
AK145087.1 | - | 3 | 585 | |||
AK172070.1 | - | 3 | 548 | |||
BC100355.1 | - | 2 | 260 | 30527188 | ||
BC117035.1 | - | 2 | 291 | |||
BC117039.1 | - | 2 | 291 | |||
BC144744.1 | - | 2 | 291 | |||
BQ444983.1 | - | 2 | 74 | |||
CA459080.1 | - | 3 | 687 |
