siRNA Details
Gene Symbol: Psmg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101430.2 | NP_001094900.1 | 3 | 431 | ||
NM_001110515.2 | NP_001103985.1 | 2 | 355 | |||
GenBank mRNA | AA063946.1 | - | 3 | 417 | ||
AA124813.1 | - | 3 | 307 | |||
AK009887.1 | - | 2 | 240 | |||
BC070417.1 | - | 3 | 405 |

More Information
Other Species Targeted

Gene Ontology Categories:
