siRNA Details
Gene Symbol: Psmg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101430.2 | NP_001094900.1 | 2 | 327 | ||
GenBank mRNA | AA063946.1 | - | 2 | 313 | ||
AA124813.1 | - | 2 | 203 | |||
BC070417.1 | - | 2 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
