siRNA Details
Gene Symbol: Hddc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027168.2 | NP_081444.1 | 2, 3 | 198 | ||
GenBank mRNA | AK009957.1 | - | 2, 3 | 139 | ||
AK161186.1 | - | 1, 2 | 1229 | |||
BC104360.1 | - | 2, 3 | 198 | 40049707 | ||
BC104361.1 | - | 2, 3 | 198 | 40049709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
