siRNA Details
Gene Symbol: Slc52a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164819.1 | NP_001158291.1 | 2 | 547 | ||
NM_001164820.1 | NP_001158292.1 | 2 | 547 | |||
NM_027172.3 | NP_081448.2 | 2 | 615 | |||
GenBank mRNA | AK009850.1 | - | 2 | 498 | ||
AK029091.1 | - | 2 | 592 | |||
AK047618.1 | - | 2 | 548 | |||
AK154970.1 | - | 2 | 597 | |||
AK170158.1 | - | 2 | 576 | |||
BC016127.1 | - | 2 | 516 | 4910227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
