siRNA Details
Gene Symbol: Ndufaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027175.3 | NP_081451.3 | 5 | 1130 | ||
GenBank mRNA | AK010328.1 | - | 4 | 960 | ||
AK050343.1 | - | 5 | 1131 | |||
BC018422.1 | - | 5 | 960 | 3593650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
