siRNA Details
Gene Symbol: Lrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081406.1 | NP_001074875.1 | 4 | 1266 | ||
GenBank mRNA | AK144741.1 | - | 4 | 1267 | ||
BC022648.1 | - | 3 | 498 | |||
BC075711.1 | - | 4 | 1248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
