siRNA Details
Gene Symbol: Ipmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027184.1 | NP_081460.1 | 6 | 847 | ||
GenBank mRNA | AK036084.1 | - | 6 | 615 | ||
AK036978.1 | - | 6 | 807 | |||
AK172246.1 | - | 6 | 939 | |||
BC052463.1 | - | 6 | 847 | 6834748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
