siRNA Details
Gene Symbol: Nkiras1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023526.3 | NP_076015.2 | 3 | 506 | ||
GenBank mRNA | AF225707.1 | - | 3 | 363 | ||
AK010285.1 | - | 4 | 679 | |||
AK028539.1 | - | 3 | 494 | |||
AK148040.1 | - | 3 | 432 | |||
AK167340.1 | - | 3 | 403 | |||
BC061092.1 | - | 3 | 411 | 6589347 |

More Information
Other Species Targeted

Gene Ontology Categories:
